Order Entry
Czech Republic
ContactUsLinkComponent
Herbicide reference standard, 3,6-Dichloropicolinic acid 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Herbicide reference standard, 3,6-Dichloropicolinic acid 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERL11690000AL
Dodavatel:  EHRENSTORFER
Číslo CAS:  
Herbicide reference standard, 3,6-Dichloropicolinic acid 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERL11690000AL
Dodavatel:  EHRENSTORFER
Číslo CAS:  

Specifikace

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    1702-17-6
  • Description:
    3,6-Dichloropicolinic acid 10 µg/ml in acetonitrile
  • Formula:
    C₆H₃Cl₂NO₂
  • Description:
    3,6-Dichloropicolinic acid

Specifikace

O tomto produktu