
4,5-Difluorosalicylaldehyde ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:199287-52-0
:
Formula:C₇H₄F₂O₂
MDL Number:MFCD08458005
:158.11 g/mol
Melting Pt:68…72 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together