Methyl-3,3-dimethoxypropionate, Sigma-Aldrich®
Catalog # SIAL288322-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:148.16 g/mol
:M-Clarity™ quality level = MQ100
EINECS:231-055-0
CAS Number:7424-91-1
Formula:C₆H₁₂O₄
MDL Number:MFCD00010650