Order Entry
Czech Republic
ContactUsLinkComponent
Zinc standard solution, 100 mg/L Zn in dil. nitric acid standard for ICP-MS
Zinc standard solution, 100 mg/L Zn in dil. nitric acid standard for ICP-MS
Catalog # ACSDM369.2NP.L1
Supplier:  CPACHEM
CAS Number:  
Zinc standard solution, 100 mg/L Zn in dil. nitric acid standard for ICP-MS
Catalog # ACSDM369.2NP.L1
Supplier:  CPACHEM
CAS Number:  

Specifications

  • Concentration:
    100 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Zinc
  • Symbol:
    Zn

Specifications

About this item