
4-Methyl-2-(trifluoromethyl)benzoic acid ≥98%
Catalog # H32534.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
About this item
CAS Number:120985-64-0
:
Formula:C₉H₇F₃O₂
MDL Number:MFCD01631591
:204.15 g/mol
:GHS07
Storage Temperature:Ambient