Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tri-tert-butylphosphine
Tri-tert-butylphosphine 10% in toluene
Tri-tert-butylphosphine 10% in toluene
ABCRS15-5811TO.50G
:
abcr
:
Tri-tert-butylphosphine 10% in toluene
ABCRS15-5811TO.50G
:
abcr
:
Pack type:
Glass bottle for solids
Pk:
50 g
:
:
202.32 g/mol
:
237-266-4
:
2845
:
4.2,I
:
13716-12-6
:
C₁₂H₂₇P
:
MFCD00015006