Order Entry
Czech Republic
ContactUsLinkComponent
Chromium standard solution, 100 mg/L Cr in dil. nitric acid standard for ICP-MS
Chromium standard solution, 100 mg/L Cr in dil. nitric acid standard for ICP-MS
Catalog # ACSDM313.2NP.L1
Supplier:  CPACHEM
CAS Number:  
Chromium standard solution, 100 mg/L Cr in dil. nitric acid standard for ICP-MS
Catalog # ACSDM313.2NP.L1
Supplier:  CPACHEM
CAS Number:  

Specifications

  • Concentration:
    100 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Chromium
  • Symbol:
    Cr

Specifications

About this item