

2,6-Dimethylpiperidin (mixture of cis- and trans-isomers) predominantly cis 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:504-03-0
:
Formula:C₇H₁₅N
MDL Number:MFCD00066304
:113.2 g/mol
Boiling Pt:126…129 °C
Melting Pt:< –20 °C
Density:0.832
Flash Pt:11 °C (51 °F)
Storage Temperature:Ambient
EINECS:207-981-6
UN:2733
ADR:3,II
:GHS02
:GHS05
Frequently Bought Together