Barium standard solution, 100 mg/L Ba in dil. nitric acid standard for ICP-MS
Catalog # ACSDM306.2NP.L1
Dodavatel: CPACHEM
Číslo CAS:
Specifikace
- Concentration:100 mg/L
- Matrix:Nitric acid dil.
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Barium
- Symbol:Ba