

2,6-Dichloropyridine-N-oxide ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2587-00-0
:
Formula:C₅H₃Cl₂NO
MDL Number:MFCD00130246
:163.99 g/mol
Melting Pt:136…140 °C
UN:2811
ADR:6.1,III
:GHS06
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together