Order Entry
Czech Republic
ContactUsLinkComponent
Chromium disilicide ≥99.5% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), Thickness: 6.35 mm (0.250 in)
Health
Chromium disilicide ≥99.5% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), Thickness: 6.35 mm (0.250 in)
Catalog # 41063.KS
CAS Number:  
Health
Chromium disilicide ≥99.5% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), Thickness: 6.35 mm (0.250 in)
Catalog # 41063.KS
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

CAS Number:12018-09-6
 :
Formula:CrSi₂
MDL Number:MFCD00168070
 :108.17 g/mol
EINECS:234-633-0
 :GHS08
Storage Temperature:Ambient