
Magnesium hexafluorosilicate hexahydrate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:18972-56-0
:
Formula:F₆MgSi·6H₂O
MDL Number:MFCD00150250
:274.47 g/mol
Density:1.79
Storage Temperature:Ambient
EINECS:241-022-2
UN:2853
ADR:6.1,III
:GHS06
Related Information
Frequently Bought Together