
3,5-Bis(trifluoromethyl)phenylhydrazine hydrochloride ≥98%
Catalog # H32975.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
About this item
CAS Number:502496-23-3
:
Formula:C₈H₇ClF₆N₂
MDL Number:MFCD03094165
:280.60 g/mol
Melting Pt:∼ 208 °C (dec.)
:GHS07
Storage Temperature:Ambient