
3-Methoxy-N,N-dimethylaniline ≥98%
Catalog # L08700.09
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
Specifications
About this item
:
:151.21 g/mol
Boiling Pt:113…114 °C (6 mmHg)
Density:1.031
Storage Temperature:Ambient
UN:2810
ADR:6.1,III
:GHS06
CAS Number:15799-79-8
Formula:C₉H₁₃NO
MDL Number:MFCD00051779