
Methyl-3-formylbenzoate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:52178-50-4
:
Formula:C₉H₈O₃
MDL Number:MFCD00189379
:164.1600 g/mol
Melting Pt:48…55 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:48 to 55 °C
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together