Order Entry
Czech Republic
ContactUsLinkComponent
4-Acetamido-2-(bis(2-hydroxyethyl)amino)anisole, Sigma-Aldrich®
4-Acetamido-2-(bis(2-hydroxyethyl)amino)anisole, Sigma-Aldrich®
Catalog # SIAL556742-25G
Supplier:  Merck
CAS Number:  
4-Acetamido-2-(bis(2-hydroxyethyl)amino)anisole, Sigma-Aldrich®
Catalog # SIAL556742-25G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ100
EINECS:246-300-7
CAS Number:24530-67-4
Formula:C₁₃H₂₀N₂O₄
MDL Number:MFCD00035865