
(Methoxymethyl)diphenylphosphine oxide ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4455-77-0
:
Formula:C₁₄H₁₅O₂P
MDL Number:MFCD00015451
:246.24 g/mol
Melting Pt:113…120 °C
EINECS:224-704-4
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together