3-Hydroxyphthalic anhydride, Sigma-Aldrich®
Catalog # SIAL308021-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:164.1150 g/mol
:M-Clarity™ quality level = MQ200
CAS Number:37418-88-5
Formula:C₈H₄O₄
MDL Number:MFCD00011557