4-Iodo-1H-pyrazole, Sigma-Aldrich®
Catalog # SIAL213993-10G
Supplier: Merck
CAS Number:
Specifications
- Typ balení:Glass bottle for solids
- Bal.:10 g
- Sekvence:3236
Specifications
About this item
:
:193.97 g/mol
:M-Clarity™ quality level = MQ200
EINECS:222-434-1
CAS Number:3469-69-0
Formula:C₃H₃IN₂
MDL Number:MFCD00005244