2-Mercaptopyridine-N-oxide sodium salt 40% in aqueous solution, Sigma-Aldrich®
Catalog # SIAL63846-500ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
Specifications
About this item
:
:149.15 g/mol
EINECS:223-296-5
CAS Number:3811-73-2
Formula:C₅H₄NNaOS
MDL Number:MFCD01941547