
Gallium indium tin alloy (62:22:16 w%), eutectic ≥99.99% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
:
MDL Number:MFCD00144388
Melting Pt:10.7 °C
Density:6.359
Storage Temperature:Ambient
UN:1760
ADR:8,III
:GHS05
Related Information
Frequently Bought Together