Order Entry
Czech Republic
ContactUsLinkComponent
3,5-Dimethylpiperidin (mixture of cis- and trans- isomers) 96%
Corrosive
Flammable
3,5-Dimethylpiperidin (mixture of cis- and trans- isomers) 96%
Certificates
undefined
Corrosive
Flammable
3,5-Dimethylpiperidin (mixture of cis- and trans- isomers) 96%

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4434 to 1.4464 (20°C, 589 nm)
  • Water:
    ≤0.5 %
  • GC:
    ≥95 %

Specification Test Results

Related Information

Frequently Bought Together