3-Methyl-1,3-pentadiene, Sigma-Aldrich®
Catalog # SIAL268976-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
ADR:3,II
:M-Clarity™ quality level = MQ100
UN:2461
CAS Number:4549-74-0
Formula:C₆H₁₀
MDL Number:MFCD00009277