Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')zinc(II) ≥99%
Supplier: abcr
Certificates
About this item
CAS Number:14363-14-5
:
Formula:C₂₂H₃₈O₄Zn
MDL Number:MFCD00192578
:431.92 g/mol
Boiling Pt:250 °C (dec.)
Melting Pt:144 °C
Frequently Bought Together