Order Entry
Czech Republic
ContactUsLinkComponent
(2,2,6,6-Tetramethyl-3,5-heptanedionato-O,O')lithium ≥98%
(2,2,6,6-Tetramethyl-3,5-heptanedionato-O,O')lithium ≥98%
Catalog # ABCRAB120843.0005
Supplier:  abcr
CAS Number:  
(2,2,6,6-Tetramethyl-3,5-heptanedionato-O,O')lithium ≥98%
Catalog # ABCRAB120843.0005
Supplier:  abcr
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

About this item

 :
 :190.24 g/mol
Boiling Pt:295 °C (dec.)
Melting Pt:265…268 °C
CAS Number:22441-13-0
Formula:C₁₁H₁₉LiO₂
MDL Number:MFCD00142754