Order Entry
Czech Republic
ContactUsLinkComponent
2,4,7,9-Tetramethyldec-5-yne-4,7-diol ((±)- and meso mixture)
2,4,7,9-Tetramethyldec-5-yne-4,7-diol ((±)- and meso mixture)
Catalog # EHERC17413600
Supplier:  EHRENSTORFER
CAS Number:  
2,4,7,9-Tetramethyldec-5-yne-4,7-diol ((±)- and meso mixture)
Catalog # EHERC17413600
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
 :226.3550 g/mol
EINECS:204-809-1
CAS Number:126-86-3
Formula:C₁₄H₂₆O₂
MDL Number:MFCD00008942