TLC plates, silica gel 60, unmodified layers, DURASIL
These unmodified standard silica layers for TLC are available as glass plates with or without fluorescent indicator.
- Hard, water-resistant and wettable layers due to a special binder system
- More polar than SIL G
Silica 60: Specific surface (BET) ~ 500 m²/g; specific pore volume 0,75 ml/g; particle size 5 to 17 μm.
Frequently Bought Together