Tubing, PVC
: MURRAY S
PVC, clear.
- Very elastic grade
- Non toxic
- Can be sterilised up to 121 °C
This tubing is very flexible and will retain its bore on bending.
Frequently Bought Together
PVC, clear.
This tubing is very flexible and will retain its bore on bending.