ContactUsLinkComponent
Eye and wound cleansing spray
  118-0247
undefined
Eye and wound cleansing spray
  118-0247
  • Description:
    Eye and wound cleansing spray, aluminium bottle filled with 150 ml sterile 0,9% sodium chloride
  • Pk:
    1
This cleansing spray is intended for rinsing and cleaning out dust and dirt from eyes and wounds. The portable spray bottle is filled with 150 ml physiological (0,9%) unbuffered sodium chloride solution.
  • Solution is preservative-free and remains sterile until last drop
  • Spray is suitable for use on both eyes and wounds
  • Actuator is protected by a plastic cap and a tamper seal
  • Nitrogen propellant is not in contact with the solution