HPLC columns, NUCLEOSIL® C18 AB
Dodavatel: MACHEREY-NAGEL
Certifikáty
O tomto produktu
NUCLEOSIL® C18 AB is a specially crosslinked octadecyl modified silica phase for HPLC.
- Robust spherical silica gel
- Strongly hydrophobic, endcapped phase with high steric selectivity for RP chromatography
- Separation mechanism based on hydrophobic and steric interactions
EC guard columns are used for analytical EC columns with the column protection system guard column holder (554-5851).
Specifikace
- Catalog No:
- MANA720936.40
- MANA720305.46
- MANA720936.46
- MANA721073.30
- MANA720935.40
- MANA720935.46
- Carbon load:
- 25%
- 25%
- 25%
- 25%
- 25%
- 25%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- pH range:
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- USP code:
- L1
- L1
- L1
- L1
- L1
- L1
Specifikace
Frequently Bought Together