ContactUsLinkComponent
VWR®, Cryoboxes for Ultra-Low Temperature Freezers
VWR®, Cryoboxes for Ultra-Low Temperature Freezers
Catalog # 479-2214
Supplier:  VWR Collection
VWR®, Cryoboxes for Ultra-Low Temperature Freezers
Catalog # 479-2214
Supplier:  VWR Collection
Specifications
  • Compartments:
    100
  • W×D×H ext.:
    133×133×50 mm
  • Seq:
    03
  • Pk:
    1
Specifications
About this item
Cryoboxes, fiberboard, sturdy, water-repellent, with covers for sample storage.
For use in ultra-low temperature freezers.
Ordering information: Box dividers available separately on request.