Vacuette® Blood Collection Sets
New Product
These safety blood collection sets are available in several versions – Either with or without Luer adapter, or with Luer-adapter and holder.
- Safety mechanism activated whilst the tip is still in the vein
- High level of protection
- Acoustic signal 'click' indicates correct activation
Especially suitable for patients with difficult vein conditions or thin veins.
Frequently Bought Together