Preparative HPLC columns, PLRP-S
: VARIAN
The PLRP-S media, rigid poly(styrene/divinylbenzene) particles, are available in a range of pore sizes for small molecule, synthetic biomolecule and macromolecule purification. Their thermal and chemical stability makes them ideal for purifications that require extreme conditions for sample preparation, compound elution and column regeneration
- Discovery stage to multi-kg cGMP production reduces method development time
- Excellent chemical stability for separations, sanitation and regeneration increases selectivity and column lifetime
- Single batch packing of multiple columns reduces system down time and validation costs
- Catalog No:
- VARIPL1712-3101
- VARIPL1712-3102
- VARIPL1712-3103
- VARIPL1512-3400
- VARIPL1512-3401
- VARIPL1712-3200
- VARIPL1512-3200
- VARIPL1512-3201
- VARIPL1512-3101
- VARIPL1512-3103
- VARIPL1712-3100
- VARIPL1212-6800
- VARIPL1512-3100
- VARIPL1212-6801
- VARIPL1512-5703
- VARIPL1212-6400
- VARIPL1212-6401
- VARIPL1512-3102
- VARIPL1512-5102
- VARIPL1512-5103
- VARIPL1512-5200
- VARIPL1212-6100
- VARIPL1212-6101
- VARIPL1212-6200
- VARIPL1512-5702
- VARIPL1212-6201
- VARIPL1512-5201
- VARIPL1512-5400
- VARIPL1812-6400
- VARIPL1512-5401
- VARIPL1812-6401
- VARIPL1812-6201
- VARIPL1812-3103
- VARIPL1712-6400
- VARIPL1212-3800
- VARIPL1212-3801
- VARIPL1812-6100
- VARIPL1712-6401
- VARIPL1712-6800
- VARIPL1812-6101
- VARIPL1812-6200
- VARIPL1712-6801
- VARIPL1512-3702
- VARIPL1712-6100
- VARIPL1212-3103
- VARIPL1712-6101
- VARIPL1512-5100
- VARIPL1812-2102
- VARIPL1712-6200
- VARIPL1212-3702
- VARIPL1812-2103
- VARIPL1212-3703
- VARIPL1812-3102
- VARIPL1712-6201
- VARIPL1512-5101
- VARIPL1512-3703
- VARIPL1212-3100
- VARIPL1212-3101
- VARIPL1212-3102
- VARIPL1712-3801
- VARIPL1712-3703
- VARIPL1712-3800
- VARIPL1712-3201
- VARIPL1212-1102
- VARIPL1712-3400
- VARIPL1712-3401
- VARIPL1212-1103
- VARIPL1712-3702
- pH range:
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- 1-14
- Temperature limit:
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- 200 ºC
- USP code:
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
- L21
Frequently Bought Together