Orders LinkContactUsLinkComponent
Capillary columns, CP-SilicaPLOT
Capillary columns, CP-SilicaPLOT
Supplier:   VARIAN
Capillary columns, CP-SilicaPLOT
Supplier:  VARIAN
Certificates
Capillary columns, CP-SilicaPLOT
Capillary columns, CP-SilicaPLOT
7 Options Available Below
Product Details & Documents
CP-SilicaPLOT brings the benefits of capillary PLOT columns (higher efficiency and faster analysis time) to many applications that previously could only be done by packed columns.
  • No influence of water on retention times for robust methodology
  • Elution of CO₂ and sulfur gases at ppm levels for improved productivity
  • Separates cyclopropane from propylene for accurate results
  • Ideal for CO₂ in ethylene, freons/CFCs, hydrocarbons, propylene and sulfur gases
  • Elutes CO₂ and sulfur gases at ppm levels and separates cyclopropane from propylene
  • Decomposition of pentadienes or CFCs is absent
Product Family Options
Product Information
  • Column Format
    Phase
    Film thickness
    Ø int.
    Length
    Availability
    Price
  • Capillary
    CP-SilicaPLOT
    4,00 µm
    0,32 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 22 956,12 Kč
    22 956,12 Kč
    1 * 1 KS
       22 956,12 Kč
  • Capillary
    CP-SilicaPLOT
    3,00 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS
  • Capillary
    CP-SilicaPLOT
    4,00 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 35 508,33 Kč
    35 508,33 Kč
    1 * 1 KS
       35 508,33 Kč
  • Capillary
    CP-SilicaPLOT
    6,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS
  • Capillary
    CP-SilicaPLOT
    6,00 µm
    0,53 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 39 385,50 Kč
    39 385,50 Kč
    1 * 1 KS
       39 385,50 Kč
  • Capillary
    CP-SilicaPLOT
    4,00 µm
    0,32 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 53 257,38 Kč
    53 257,38 Kč
    1 * 1 KS
       53 257,38 Kč
  • Capillary
    CP-SilicaPLOT
    6,00 µm
    0,53 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 54 546,36 Kč
    54 546,36 Kč
    1 * 1 KS
       54 546,36 Kč