Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Chemicals955-590.jpg
Solvents, HPLC Grade, VWR Chemicals BDH
Manufactured to the most stringent requirements both in Europe and the USA, this portfolio of solvents and modifiers provides outstanding value for the money.
Solvents, HPLC Grade, VWR Chemicals BDH​ | Avantor
These solvents:
  • Are designed to meet demanding HPLC and UV/Vis analytical and quality control requirements
  • Are filtered down to 0.2µm level and bottled under nitrogen ‒ Feature tamper-evident caps for added peace of mind
  • Carry Globally Harmonized System (GHS) labeling to help with compliance ‒ Are available in glass bottles that are specially treated to ensure low conductivity and long shelf life
  • Feature QR codes on labels for quick access to SDS and C of A
  • Feature environmentally-friendly corrugated/pulp case inserts that are fully recyclable
  • Are also available in 200L VWR ReCAP returnable containers, for all grades of Acetonitrile and Methanol
View our range of most popular Solvents, HPLC Grade.