Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
thermo_environmental_containers_900.jpg
Quality Containers for Environmental Sample Collection, Transport and Preservation
Thermo Scientific Environmental Sample Containers are produced in our ISO 9001 registered manufacturing facilities in the USA.
Thermo Scientific Environmental Containers | Avantor
All of our processes – from design to development to manufacturing – meet or exceed the requirements for quality as set forth by the International Standards Organization.
In addition, we have developed Thermo Scientific Performance-based Specifications, which guarantee our processed and certified products meet or exceed US EPA analyte specifications as required in Section II.B of the current “Specifications and Guidance for Contaminant-free Sample Containers” document.
Product selection
thermo_volatile_organic_analysis_100.jpg
Our range of VOA containers is suited for collection, transport and analysis. Featuring vials with septa and solid closures for any application.
thermo_semivolatile_containers_100.jpg
Rely on our clear or amber glass containers for your important sample collection. Available Certified to meet EPA Performance-based Specifications for semi-volatile organics, pesticides, PCBs and metals anlysis
thermo_metals_water_quality_containers_100.jpg
Containers recommended for critical projects where sample containment and transport is of elevated concern. Available Certified to meet EPA Performance Based Specifications for semi-volatile organics, pesticides, PCBs and metals analysis
thermo_bulk_hdpe_separates_containers_100.jpg
Offering all the containers you need from certified clean containers to unassembled containers and caps ready for your own preparation procedures