Order Entry
Czech Republic
ContactUsLinkComponent
asecos1920-760.png
asecos - Your expert for hazardous materials storage and handling
Safe storage of hazardous materials with asecos
Have you ever thought about the effects of a fire involving flammable liquids?
As a leading manufacturer of safety storage cabinets in accordance with EN 14470-1/2 for the storage of hazardous substances in the workplace, asecos is an expert in this field. In our fire test,we illustrate the importance of fire resistance using the example of three safety storage cabinets. With a type 90 safety storage cabinet from asecos, you gain valuable time for evacuation in the event of a fire.
The innovative and high-quality safety solutions from asecos, made in Germany, not only guarantee safety in the workplace, but also stand for responsibility and sustainability.
Literature
Main-catalog400-495.png
Safe working with hazardous substances.
asecos-ION-LINE400-495.png
Lithium-ion cabinets as a safe solution for the active and passive storage of lithium batteries.
asecos-waste-disposal400-495.png
Underbench cabinets for the safe collection and disposal of hazardous substances.
asecos-V-LINE400-495.png
Vertical cabinets for the safe storage of hazardous substances.
asecos V-CLASSIC400-495.png
Innovative and sustainable vertical cabinets for the safe storage of hazardous substances.
Videos
Product selection
Q-Line400-495.png
The all-rounder among fire-protected safety storage cabinets. They are suitable for storing flammable hazardous materials in work areas.
UB-Line400-495.png
The certified complete system ensures that solvent waste is disposed of safely and in accordance with regulations. The type 90 base cabinet is electrically conductive and equipped with high-quality components.
SL-Line400-495.png
The acid/alkali cabinets are suitable for the safe storage of aggressive acids and alkalis. The separate storage chambers offer a safe and efficient storage option.
UFA-Line400-495.png
With the plug-in recirculating air filter attachment, solvent vapors in safety cabinets can be permanently retained until the filter is saturated.
V-Line400-495.png
The V-LINE MULTIRISK is the solution for safe, flexible and energy-efficient storage of hazardous materials. This cabinet enables space-saving and unrestricted storage of different hazardous substances.
ION-Line400-495.png
The ION-LINE lithium cabinets offer the right protection for the safe charging or storage of lithium batteries. With three different safety levels, you will find the right lithium cabinet for you.