Order Entry
Czech Republic
ContactUsLinkComponent
heroimagesamplepreparaton.jpg
Sample Preparation
Sample preparation can improve the result of an analytical method, e.g., pre-concentration of targeted analytes, and/or minimize the background interferences from the sample matrix. Avantor® has an extensive range of chemicals, consumables and equipment to help you achieve success with a high-quality and reliable sample preparation process.
Products
1stproductsamplepreparation.jpg
Specific polypropylene vials recommended when analysing polar compounds /analytes and dimensionally verified/inspected by automated optical and gauges to ensure critical dimensions and tolerances.
2stproductsamplepreparation.jpg
Special phase for SPE analysis. For enrichment of PFAS from water, textiles and sediments (contaminated soils). Polymeric combination phase with weak anion exchanger, highly porous, spherical particles.
3stproductsamplepreparation.jpg
Wide choice of caps which can be used with all HPLC/UHPLC instrument types.
4stproductsamplepreparation.jpg
Optimize the flow path in your chromatography lab with your choice of capillaries, fittings and connectors.