ContactUsLinkComponent
[EN]STERILE PARAFILM 101 X 152 MM 1 * 5 KS
AMCOR Parafilm® Sterile, Strips
Catalog # HEAT234526BSTL
Supplier:  HEATHROW
New Product
undefined
AMCOR Parafilm® Sterile, Strips
Catalog # HEAT234526BSTL
Supplier:  HEATHROW
New Product
Specifications
  • Sterilní:
    +
  • Barva:
    Přírodní
  • Popis:
    Parafilm® strips, dimension: 101×152 mm
  • Sekvence:
    2
Specifications
About this item
Sterile strips of semi-transparent, flexible, thermoplastic film with two-way stretch.
  • Packaged and sterilised in USA
  • Sterile expiration: Two years
  • Non-toxic, odourless, and semi-transparent
  • Creates a tight moisture barrier that holds a strong seal and doesn’t release from glassware
  • Prevents moisture and volume loss due to material composition and tight seal
  • The flexible and self-sealing material allows it to cling around irregular shapes and surfaces
  • Resistant up to 48 hours against saline solutions, inorganic acids, and alkaline solutions
  • Sterile
Ideal for protecting samples and preventing contamination or evaporation - perfect for cleanroom environments and strict sanitation protocols.
These pre-cut strips are sterilised using a vapour hydrogen peroxide process, adding an extra layer of protection to Parafilm’s natural waterproof and moisture-resistant properties.
Parafilm® is a flexible, semi-transparent thermoplastic film designed for reliable sealing in laboratory, medical, and industrial environments. Engineered from a unique blend of waxes and polyolefins, Parafilm® stretches up to 200% of its original size, conforming tightly to irregular surfaces such as test tubes, beakers, petri dishes, and flasks. Each strip comes individually packaged in a sealed, lot-traceable pouch featuring a colour sterilisation indicator - ensuring both safety and convenience.
Frequently Bought Together