[EN]MM 1-43FX 5X100 UG 1 * 100 µG
Katalogové číslo AATB21484
Dodavatel: AAT Bioquest
New Product
Frequently Bought Together
Aceton ≥99%, TECHNICAL
Dodavatel: VWR Chemicals
Voda, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
Kyselina trifluoroctová, Uvasol® for spectroscopy, Supelco®
Dodavatel: Merck
Pipette tips, Finntip™
Dodavatel: Thermo Fisher Scientific
Ethanol 96% vol Ph. Eur.
Dodavatel: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Dodavatel: VWR Chemicals
Ethanol absolute ≥99.5% USP, Ph. Eur.
Dodavatel: VWR Chemicals
Voda, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Dodavatel: VWR Chemicals
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
Kyvety jednorázové
Dodavatel: Brand
VWR®, Pipety Pasteurovy, High performance
Dodavatel: VWR Collection
Aceton ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals