[EN]P100-LMARK-PTFE PLUNGER PK2 1 * 2 KS
SGEAKLMK.26367072P
New Product
[EN]P100-LMARK-PTFE PLUNGER PK2 1 * 2 KS
SGEAKLMK.26367072P
New Product
Frequently Bought Together
HPLC columns, LiChrospher®
: Merck
VWR®, Screw Caps for Screw Neck Vials ND13
: VWR Collection
Avantor® ACE® C18, HPLC Columns, 5 µm
: Avantor
Sorbents, Superspher®
: Merck
Avantor® ACE®, Guard Cartridge Holders
: Avantor
VWR®, Screw Caps for Short Thread Vials ND9
: VWR Collection
SafetyWasteCaps
: S.C.A.T.
Silikagel 60, irregular 40 - 63 µm for flash chromatography
: VWR Chemicals
Inlet liners
: Agilent
Screw caps for 9 mm screw neck vials, Chromacol™
: CHROMACOL
VWR®, Snap Caps for Snap Ring Vials ND11
: VWR Collection
Inserts for screw neck vials, N 8 and crimp neck vials, N 11
: MACHEREY-NAGEL
Caps for 20 ml glass crimp top headspace vials, level 1 everyday analysis, SureSTART™
: Thermo Fisher Scientific
SCV gas sampling bags, Tedlar®
: Merck
Crimpers and decappers, WHEATON®
: DWK Life Sciences
SureSTART™ Glass Inserts for 2 ml Vials, Level 3 High Performance Applications
: Thermo Fisher Scientific