ContactUsLinkComponent
Floating Vial Rack, 20 mm, Black
  HEAT121006
 :  HEATHROW
New Product
undefined
Floating Vial Rack, 20 mm, Black
  HEAT121006
 :  HEATHROW
New Product
  • Barva:
    Černá
  • Popis:
    Floating vial rack, dimensions: 127×86×64 mm, quantity: pack of 5
  • Materiál:
    HDPE
Securely holds vials in a water bath or any liquid environment.
  • Detachable carrying handle for easy lifting
  • Designed for any vial with 20 mm diameter and AT-Vials up to 50 ml
  • Made of high-density and water-resistant polyethylene
  • Temperature range: 0 to 70 °C
  • Handle length: 64 mm
  • Array: 3×2
  • No. of wells: 6
  • Well diameter: 20 mm
Designed with high-density foam, this floating rack is buoyant, durable, and chemical-resistant, ensuring that your samples remain stable and organised during incubation or other liquid-based experiments.
Frequently Bought Together