[EN]MONKEY BETA AMYLOID 42 ELISA KIT 1 * 96 Tests
Katalogové číslo ANTIA326423-96
Dodavatel: ANTIBODIES.COM
New Product
[EN]MONKEY BETA AMYLOID 42 ELISA KIT 1 * 96 Tests
Katalogové číslo ANTIA326423-96
Dodavatel: ANTIBODIES.COM
New Product
Frequently Bought Together
Aceton ≥99%, TECHNICAL
Dodavatel: VWR Chemicals
Sealing film for manual application, Parafilm® M
Dodavatel: HEATHROW
VWR®, Ubrousky čisticí
Dodavatel: VWR Collection
VWR®, Nitrile Gloves
Dodavatel: VWR Collection
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Dodavatel: VWR Chemicals
Voda, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
VWR®, Sérologická pipeta, standardní
Dodavatel: VWR Collection
Rukavice, nitrilové, Kimtech™ Purple Nitrile™
Dodavatel: KIMBERLY CLARK
VWR® NITRILE LIGHT, Rukavice, nitrilové
Dodavatel: VWR Collection
VWR®, Pipety Pasteurovy, High performance
Dodavatel: VWR Collection