CytoFLEX Mosaic Detection Modules
8, CytoFLEX LX and CytoFLEX Mosaic 88 spectral flow analyzer system U-V-B-Y-R (5 laser UV)
Katalogové číslo BECMD20161
New Product
Dodavatel: Beckman Coulter
Přejít na:
Product Details & Documents
The CytoFLEX Mosaic Detection Module is the natural evolution of the CytoFLEX platform. A smart approach to spectral analysis, the detection module connects to the CytoFLEX LX/S flow cytometer, providing the flexibility to mix and match different detection methods in one instrument.
A flexible approach to spectral flow cytometry
With up to 88 detection channels, the CytoFLEX Mosaic Detection Module enhances the CytoFLEX S or LX flow cytometer, empowering users to tackle the most complex experiments. Together, they create the most advanced and powerful CytoFLEX flow cytometry solution to date.
Specifikace
- Popis:CytoFLEX LX and CytoFLEX Mosaic 88 spectral flow analyzer system U-V-B-Y-R (5 laser UV)
- Sekvence:8