[EN]LABEL CST-29NPNOT0.5C1-1WH 1000/ROLL 1 * 1 Roll
Katalogové číslo GAIN29NPNOT05C11WH
Dodavatel: GA
New Product
[EN]LABEL CST-29NPNOT0.5C1-1WH 1000/ROLL 1 * 1 Roll
Katalogové číslo GAIN29NPNOT05C11WH
Dodavatel: GA
New Product
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Dodavatel: VWR Chemicals
Voda, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
Aceton ≥99.5%, GPR RECTAPUR®
Dodavatel: VWR Chemicals
Aceton ≥99%, TECHNICAL
Dodavatel: VWR Chemicals
VWR®, Ubrousky čisticí
Dodavatel: VWR Collection
VWR®, Nitrile Gloves
Dodavatel: VWR Collection
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Dodavatel: VWR Chemicals
VWR®, Pipety Pasteurovy, High performance
Dodavatel: VWR Collection
VWR®, Sérologická pipeta, standardní
Dodavatel: VWR Collection
Pipette tips, Finntip™
Dodavatel: Thermo Fisher Scientific
Voda, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Dodavatel: VWR Chemicals
Příslušenství pro Controlled Rate Freeze Thaw, CRFT
Dodavatel: GRANT INSTRUMENTS