[EN]SWABCAP VALVE CAP WITH 70% IPA 1 * 200 KS
Katalogové číslo BRAUEM-SCXT3
Dodavatel: B. Braun
New Product
[EN]SWABCAP VALVE CAP WITH 70% IPA 1 * 200 KS
Katalogové číslo BRAUEM-SCXT3
Dodavatel: B. Braun
New Product
Frequently Bought Together
Aceton ≥99%, TECHNICAL
Dodavatel: VWR Chemicals
Pipette tips, epT.I.P.S.® Standard (General Lab Product)
Dodavatel: EPPENDORF
VWR®, Nitrile Gloves
Dodavatel: VWR Collection
VWR®, Ubrousky čisticí
Dodavatel: VWR Collection
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Dodavatel: VWR Chemicals
Voda, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Dodavatel: VWR Chemicals
VWR®, Sérologická pipeta, standardní
Dodavatel: VWR Collection
Pipette tips, Finntip™
Dodavatel: Thermo Fisher Scientific
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Dodavatel: VWR Chemicals
Voda, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
VWR®, Pipety Pasteurovy, High performance
Dodavatel: VWR Collection
Extension and connection tubing
Dodavatel: B. Braun
Chlorid sodný, EMSURE® ACS, ISO, Reag. Ph. Eur. p.a., Supelco®
Dodavatel: Merck