[EN]MICROSLIDE WHITE SNOWCOAT GRND CLPD 1 * 1.440 KS
Katalogové číslo LEIH3808100
Dodavatel: LEICA HISTOLOGY
New Product
[EN]MICROSLIDE WHITE SNOWCOAT GRND CLPD 1 * 1.440 KS
Katalogové číslo LEIH3808100
Dodavatel: LEICA HISTOLOGY
New Product
Frequently Bought Together
Aceton ≥99%, TECHNICAL
Dodavatel: VWR Chemicals
VWR®, Pipety Pasteurovy, High performance
Dodavatel: VWR Collection
Aceton ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Dodavatel: VWR Chemicals
Acetonitril, LiChrosolv®, hypergrade for LC-MS, Supelco®
Dodavatel: Merck
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Dodavatel: VWR Chemicals
Voda, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
Ethanol absolute ≥99.5% USP, Ph. Eur.
Dodavatel: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Dodavatel: VWR Chemicals
Pipette tips, Finntip™
Dodavatel: Thermo Fisher Scientific
VWR®, Nitrile Gloves
Dodavatel: VWR Collection
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Dodavatel: VWR Chemicals
Decontamination solution, DNA AWAY®
Dodavatel: Thermo Fisher Scientific
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals