b.safe Laboratory Flasks
01, 500 ml, GL 45
Katalogové číslo BOHLR105-79
Dodavatel: Bohlender
Přejít na:
Product Details & Documents
Laboratory flask made of borosilicate glass.
Includes screw cap and spout made of PP
Specifikace
- Závit:GL 45
- Kapacita:500 ml
- Sekvence:01