Orders LinkContactUsLinkComponent
Label Sheets, Core2Write
Label Sheets, Core2Write
04, Core2Write label, sterile, irradiated, Bílá, 101,6×50,8 mm, 10 labels/sheet, 500 sheets/case
Katalogové číslo VELTC2WR4X2SHEET02
Dodavatel:   VELTEK ASSOCIATES
Label Sheets, Core2Write
Katalogové číslo VELTC2WR4X2SHEET02
Dodavatel:  VELTEK ASSOCIATES
Label Sheets, Core2Write
Label Sheets, Core2Write
04, Core2Write label, sterile, irradiated, Bílá, 101,6×50,8 mm, 10 labels/sheet, 500 sheets/case
Přejít na:
Price
119 790,00 Kč
Přihlásit se pro dostupnost
1 * 5.000 KS
Product Details & Documents
Constructed using CleanPrint 10, Core2Write labels are extremely durable, smear and smudge proof, chemical resistant, and remain pliable in extreme temperatures.
  • Can be applied to sub-zero surfaces
  • Perforation between labels is available
All labels can be customised or left blank.
Informace pro objednávání: Also available in sheets (sold separately).
Specifikace
  • Barva:
    Bílá
  • Popis:
    Core2Write label, sterile, irradiated
  • Rozměry:
    101,6×50,8 mm
  • Sekvence:
    04
  • Balené:
    10 labels/sheet, 500 sheets/case