Orders LinkContactUsLinkComponent
Label Sheets, Core2Write
Label Sheets, Core2Write
01, Core2Write label, non-sterile, Bílá, 50,8×25,4 mm, 10 labels/sheet/500 sheets/case
Katalogové číslo VELTC2WR2X1SHEET01
Dodavatel:   VELTEK ASSOCIATES
Label Sheets, Core2Write
Katalogové číslo VELTC2WR2X1SHEET01
Dodavatel:  VELTEK ASSOCIATES
Label Sheets, Core2Write
Label Sheets, Core2Write
01, Core2Write label, non-sterile, Bílá, 50,8×25,4 mm, 10 labels/sheet/500 sheets/case
Přejít na:
Price
119 790,00 Kč
Přihlásit se pro dostupnost
1 * 18.000 KS
Product Details & Documents
Constructed using CleanPrint 10, Core2Write labels are extremely durable, smear and smudge proof, chemical resistant, and remain pliable in extreme temperatures.
  • Can be applied to sub-zero surfaces
  • Perforation between labels is available
All labels can be customised or left blank.
Informace pro objednávání: Also available in sheets (sold separately).
Specifikace
  • Barva:
    Bílá
  • Popis:
    Core2Write label, non-sterile
  • Rozměry:
    50,8×25,4 mm
  • Sekvence:
    01
  • Balené:
    10 labels/sheet/500 sheets/case